Learn More
Juchheim Laborgeraete™ Perforated Coverplate for Laboratory Fire Suppression Trough
Product Code. 10265062
Click to view available options
Color:
Gray
Unit Size:
Each
Product Code. 10265062
Brand: Juchheim Laborgeraete™ 00311
Only null left
Add to basket
Add to basket
Accessories for stainless steel fire protection tray. Suitable for fire protection tray No.00310.
  • Distributes the weight of parked containers
  • Improves fire extinguishing properties
    .
  • Made of stainless steel
Specifications
Type Bin Lid
Material Stainless Steel
Color Gray
Shape Square
Length (Metric) 450 mm
Width (Metric) 450 mm