Learn More
Juchheim Laborgeraete™ Perforated Coverplate for Laboratory Fire Suppression Trough
Click to view available options
Color:
Gray
Unit Size:
Each
Description
- Distributes the weight of parked containers
- Improves fire extinguishing properties
. - Made of stainless steel
Specifications
Specifications
| Type | Bin Lid |
| Material | Stainless Steel |
| Color | Gray |
| Shape | Square |
| Length (Metric) | 450 mm |
| Width (Metric) | 450 mm |