missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for Glass Microanalysis Filter Holders
Accessories for filter holders for sample preparation.
Brand: Merck XX1012508
211.52 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.80530kg
Description
Accessories for filter holders for sample preparation.
Specifications
Not Autoclavable | |
25 mm glass microanalysis vacuum filter | |
Non-sterile |
Base with No. 5 stopper | |
Silicone | |
Perforated Stopper |