missing translation for 'onlineSavingsMsg'
Learn More

Peri-xanthenoxanthene, 95%, Thermo Scientific™

Catalog Number p-7077518
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
191-28-6
C20H10O2
282.3
AMDQVKPUZIXQFC-UHFFFAOYSA-N
dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
240798
C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4
This item is not returnable. View return policy
191-28-6
C20H10O2
282.3
AMDQVKPUZIXQFC-UHFFFAOYSA-N
dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
240798
C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 282.3
Percent Purity 95%
Chemical Name or Material Peri-xanthenoxanthene
compliance-icons
Product Identifier
  • Peri-xanthenoxanthene, 95%
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'