Learn More
Macherey-Nagel™ Permabond™ SE-52 GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.25 μm
0.5 μm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.5 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | Permabond™ SE-52 |
| USP Type | G27, G36 |
| For Use With (Application) | Gas Chromatography (GC) |
| Temperature Range | 300°C to 320°C |
| Film Thickness | 0.5 μm |
| Length (Metric) | 25 m |
| Packing Material | Permabond™ SE-52 |
| Show More |