missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Peroxtesmo KO Qualitative Test Paper

Product Code. 11366504 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Blue/Green
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11366504

Brand: Macherey-Nagel™ 90606

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Peroxtesmo KO allows the quick and easy detection of peroxidase in food. It is used in the food processing industry to evaluate the quality of canned food.

Specifications

Color Indicator White to Blue/Green
For Use With (Application) For detection of Peroxidase in milk
Quantity 100 Pack
Test Time 2 min.
Type Qualitative Test Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.