missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Kit, SPEX CertiPrep™

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Kits containing 10-multi-compound mixes are ideal for pesticide screening, food safety and QuEChERS applications.

Brand:  SPEX Certiprep SPXPR-KIT

Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II

Product Code. 16370488

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Pesticide Kit containing 10-multi-compound mixes
  • A total of 144 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Services

1 year

Specifications

Specifications

Trifloxystrobin
Boscalid
Fenoxycarb
Piperonyl butoxide
Tebufenpyrad
Iprodione
Imid
100μg/mL
Glass Ampule
99.5
99.5
99
95.8
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
97.6
99.2
99.3
99.5
98.5
99.4
98.8
99.4
99.5
99.2
99.5
98.3
99.5
99.4
99.3
99.5
99.5
99.5
97.4
99.9
98.6
98.4
98.9
99.5
99.6
99.5
99.5
98.5
99
96
98.9
99
99.4
99.1
98.8
97.5
99.5
98
99
99.6
99.5
99.5
99.2
99.2
99.5
98
99.5
98
98.2
99.5
99.5
99.9
99.9
99.5
98.6
99.2
99.5
99.5
99.5
99.5
98.4
98.8
99.1
99.5
95
99.7
99.2
99.4
99.5
99.5
98.4
99.4
97.5
99.5
98.9
99
99.5
98.4
99
98
98.2
97.7
98.1
99.4
25
99.3
99
99.5
99.5
99.5
98
97.5
99.5
99
98.7
99.5
99.6
99.4
98.6
99.5
97
97
99.5
99.5
96.7
94.2
97
99.5
98.1
98
98.4
98
99.5
99.5
98.6
99.4
98.4
99
C20H19F3N2O4
C18H12Cl2N2O
C17H19NO4
C19H30O5
C18H24ClN
10 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions