missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss. p.a., Reag. Ph. Eur., Honeywell™

Catalog Number p-7073764
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
101316-46-5
MFCD00081849
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
101316-46-5
MFCD00081849
UN Number 1268
Grade puriss. p.a.
Density 0.661 to 0.664g/mL at 20°C
Quantity 1 L
Flash Point −26°C (−14.8°F)
Chemical Name or Material Petroleum Ether, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'