Learn More
Cole-Parmer™ PFA Compression Threaded Adapter
Click to view available options
Tubing Size:
10mm OD
Unit Size:
Each
Description
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
| Description | Compression x NPT(M) |
| Material | PFA |
| Tubing Size | 10mm OD |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Thread Size | 3/8 in. |
| Product Type | PFA compression threaded adapter, 10mm tubing OD x 3/8in male NPT |