Learn More
Schmidlin™ PG Plus FID Station H2 and Zero Air Generator
Product Code. p-7228510
Encompass_Preferred
Click to view available options
Flow Rate:
100 mL/min.
250 mL/min.
Wattage:
65 W
115 W
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17748879
Brand: Schmidlin™ DBFHG100EU
Regulatory: This product is under eco-contribution tax.
This dual gas generator produces a continuous supply of high purity hydrogen and zero air.
  • Increase efficiency and improve performance
  • On-demand supply 24/7
  • H2 flow rate and zero air up to 5 L/min.
  • THC Pressure: 11 barg (160 psig)
  • Permeation membrane drying system
  • USB connectivity
  • Easy to install with 'plug and play' functionality
  • Ideal for all GC detector applications
Specifications
Depth (Metric) 68 cm
Flow Rate 100 mL/min.
Height (Metric) 15 cm
Max. Ambient Operating Temperature 35°C
Outlet Pressure 11 bar
Width (English) 27 in.
Width (Metric) 69 cm
Connections Outlet: 1/8 in.
Electrical Requirements 110/120 V at 60 Hz, 220/240 V at 50 Hz
Nitrogen Purity >99.9996%
Wattage 65 W