missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 12.450 Millesimal Solution Buffers, Hanna Instruments™
Click to view available options
Quantity:
500 mL
Description
- Premium quality pH 12.450 millesimal calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
- Designed for customers who require a resolution of 0.001 for their pH measurements
- Supplied with Certificate of Analysis
- Accuracy of +/- 0.002 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each bottle
- Produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
Specifications
Quantity | 500 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'