missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 5.00 Technical Calibration Buffer Sachets, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Unit Size:
Pack of 25
Description
- Premium quality pH 5.00 technical calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
- Supplied with Certificate of Analysis
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- Temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
| Color | Undesignated |
| Concentration | >1 to <10% |
| pH | 5.00 |
| Physical Form | Liquid |
| Quantity | 20 mL |
| Formulation | Citric acid monohydrate 2 |
| Odor | Odorless |
| Packaging | 25Pack |
| Traceability to NIST | Yes |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction