missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 7.01 Technical Calibration Buffer Sachets, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Unit Size:
Each
Description
- A premium quality pH 7.01 technical calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Technical calibration buffer produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
| pH | 7.00 |
| Quantity | 20 mL |
| Packaging | 25Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction