missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 7.00, Colored, Chem-Lab

Product Code. 11953073
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11953073

Brand: Chem-Lab CL03.0704.0100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for pH maintenance and measurement in neutral samples. Buffer Solution, pH 7.00, Colored, Chem-Lab is a green phosphate solution with a pH level of 7.00 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.

pH Level ± 0.02 at specified temperature:

  • pH 7.13 at 0°C
  • pH 7.05 at 10°C
  • pH 7.00 at 20°C
  • pH 6.99 at 25°C
  • pH 6.98 at 30°C
  • pH 6.97 at 40°C
  • pH 6.96 at 50°C

Specifications

Boiling Point 100°C
Color Green
Density 1.00g/mL
For Use With (Application) Laboratory use, pH measurements
pH 7.00
Physical Form Liquid
Quantity 100 mL
Solution Type pH Buffer
Formulation Phosphate
Odor Odorless
Packaging Polyethylene
Traceability to NIST Yes
Solubility Information Soluble in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.