Learn More
Knick Elektronische™ pH meter Portavo 904X digital Atex
Click to view available options
Calibration Points (pH):
1-, 2-, and 3-point
Unit Size:
Each
Description
- Basic unit for mobile pH measurement for Atex areas (1 and 2.)
- Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
- Large easy-to-read LC display with 15mm character height is constructed of antireflection natural glass, scratchproof and resistant to chemicals. Clear icons for operator guidance.
- Integrated, replaceable quiver protects the sensor. Transparent. Easy to remove.
- Blue, ergonomic enclosure is resistant to impact and chemicals.
- Water-tight, diecast chassis (IP 66 protection.)
- Cover protects against dirt and damage.
- Highly flexible lamellar joint can be folded back completely.
- Fold-out hook for hanging or standing up keeps both hands free for handling the sensor.
- Robust, gold-plated water-tight sockets.
- Carrying strap fixed directly to the meter.
- 1-, 2-, and 3-point calibration.
- Calimatic™ automatically recognizes the right buffer from 8 buffer sets.
- Manual calibration possible with any two buffers.
Specifications
Specifications
| Includes | Batteries, Electrode Quiver, and 911 x pH Carrying Strap, without Electrode |
| For Use With (Application) | Mobile pH measurement for Atex areas (1 and 2) |
| Calibration Points (pH) | 1-, 2-, and 3-point |
| Display Type | LCD |
| Keypad | Smooth membrane keypad |
| Model | 904X Atex |
| Type | Mobile pH Meter |