Learn More
WTW™ pH/mV Module ADA 94pH/IDS with BNC and 4mm Banana Plug
Description
WTW offers adapter solutions for mobile energy supply, data transfer of old systems as well as for the connection of conventional pH electrodes to digital systems. The USB adapters ADA USB/Ser and ADA USB are used to connect devices with RS 232 interface to PCs and laptops with USB interfaces, especially all lab and portable meters with RS 232 connector (portable meters 3xxi, benchtop meters inoLab™ Level 2, 730, field meters 197i, 1970i).
Specifications
Specifications
| Type | pH/mV Module |
| Includes | Mounting Accessories |
| For Use With (Equipment) | InoLab Multi 9420/9430 IDS, for pH/ORP/ISE Electrodes |