Learn More
Hanna Instruments™ pH/ORP/Temperature Benchtop Meter
Product Code. 11348610
Click to view available options
Dimensions (L x W x H):
182 L x 240 D x 74mm H
Unit Size:
Each
Product Code. 11348610
Brand: Hanna Instruments™ HI222102
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.01
Calibration Up to 5 point calibration, 7 standard buffers available
Calibration Points Up to 5 point calibration, 7 standard buffers available
Data Points 100
Detection Range -2 to 16.00°C
Display Type LCD
Range (mV) ±699.9 mV, ±2000 mV
Range (pH) -2 to 16
Resolution (pH) 0.01
Temperature Range (Metric) -20.0°C to +120.0°C
Inputs BNC
Model HI2221-02
Power Supply 230V
Probe Type HI 1131P glass body, single junction, refillable with BNC connection and 1 m (3.3') Cable HI 7662 stainless steel probe
Product Type pH/ORP/Temperature Benchtop Meter
Resolution (mV) 0.1mV
Temperature Accuracy ±0.2°C
Temperature Compensation Automatic or manual from –20 to 120°C
Temperature (Metric) Operating -20°C to +120.0°C
Temperature Probe Calibration HI 1131P glass body, single junction, refillable with BNC connection and 1 m (3.3') Cable HI 7662 stainless steel probe
Temperature Resolution 0.1°C
Dimensions (L x W x H) 182 L x 240 D x 74mm H
Width (Metric) 240 mm
Height (Metric) 74 mm
Depth (Metric) 182 mm
Electrical Requirements 240V, 50/60Hz
Voltage 240 V
Frequency 50Hz