Learn More
Cowie Technology™ PTFE Joint Adapter
Product Code. 10363452 Shop All Fisherbrand Products
Change view
Click to view available options
Standard Taper Joint No:
NS 10/19, NS 19/26
NS 14/23, NS 19/26
NS 19/26, NS 29/32
NS 19/26, NS 34/35
NS 24/29, NS 29/32
NS 24/29, NS 34/35
NS 29/32, NS 34/35
Cone Size:
19/26
29/32
34/35
Joint Size:
10/19
14/23
19/26
24/29
29/32
Unit Size:
Each
Product Code. Standard Taper Joint No Cone Size Joint Size unitSize
10363452 NS 14/23, NS 19/26 19/26 14/23 Each
10211762 NS 10/19, NS 19/26 19/26 10/19 Each
10271852 NS 19/26, NS 34/35 34/35 19/26 Each
10313602 NS 19/26, NS 29/32 29/32 19/26 Each
10363692 NS 24/29, NS 29/32 29/32 24/29 Each
10150382 NS 29/32, NS 34/35 34/35 29/32 Each
10465252 NS 24/29, NS 34/35 34/35 24/29 Each
7 options
Product Code. 10363452
Only null left
Add to basket
Add to basket
Cowie Technology™ PTFE Joint Adapters are grease-free and create high vacuum seals for standard taper ground glass joints.
  • Grease-free
  • High vacuum seals for standard taper ground glass joints
  • Substantial shoulder for ease of handling and repeated use
  • High vacuum performance (leakage rate is less than 1 x 10-4 toor, litre, sec.-1) is attained using transverse sealing rings.
  • Use up to 280°C
Specifications
Quantity 1 Each
Type PTFE Joint Adapters
Cone Size 19/26
Joint Size 14/23
Standard Taper Joint No NS 14/23, NS 19/26