missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PHA 767491 hydrochloride

Description
ATP-competitive cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 0.034 - 0.75, 0.24 - 1.2, 0.25 - 1.1 and 0.46 - 1 μM for cdk9, cdk2, cdk1 and cdk5, respectively). Also inhibits mitogen-activated protein kinase-activated protein kinase-2 (MK2; IC50 = 171 nM). Prevents initiation of DNA replication. Inhibits cell proliferation in a variety of human cell lines and induces apoptosis in a p53-independent manner in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 249.7 |
| Purity | >99% |
| Molecular Weight (g/mol) | 249.698 |
| Chemical Name or Material | PHA 767491 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?