Learn More
Heidolph instruments PharMed™ 2-Stop Tubing
Ideal tubing for medical, laboratory, and research uses. Heidolph™ PharMed™ 2-Stop Tubing is durable, non-toxic and designed to prevent slipping while the pump is in operation.
Brand:  Heidolph instruments 525-20013-00
Product Code. 10751483
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • High fatigue strength under repeated reversed bending stresses.
  • Non-toxic, biocompatible.
  • Very low gas permeability.
  • Well suited to acids and bases.
  • Designed with 2 stops to prevent slipping while the pump is operating.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDA (21 CFR 177.2600), USP Class VI, GLP, Pharmacopoea and Europaea
0.51 mm
2.31 mm
High Fatigue Strength, Non-Toxic, Biocompatible, Very Low Gas Permeability, Well-Suited to Acids and Bases.
Non-sterile
Autoclave, Ethylene Oxide, Irradiation
2 stops
Additives may migrate.
Autoclavable
Thermoplastic
Pump
40°C
0.51 mm I.D./2.31 mm O.D
7.0 m Water
Very low