Learn More
Olympus™ Phase Contrast Ring, Small
Provide phase contrast viewing with this small, insertable ring. Olympus™ Phase Contrast Ring, Small works with the U-UCD8 universal condenser, and a variety of applicable objectives.
Brand: Olympus™ K10017625
Additional Details : Weight : 0.01000kg
Description
- Provides phase contrast viewing.
- Small.
- Insertable.
- Works with U-UCD8 universal condenser.
Specifications
Phase Contrast Ring, Small | |
PlanN40XPh, UPlanFLN40XPh |