Learn More
Olympus™ Phase Contrast Ring, Small
Provide phase contrast viewing with this small, insertable ring. Olympus™ Phase Contrast Ring, Small works with the U-UCD8 universal condenser, and a variety of applicable objectives.
Brand:  Olympus™ K10017626
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12661561
Quantity Price
1 211.00€ / Each
6 172.53€ / Each
Estimated Shipment
16-04-2021
Add to basket
Description
Description
  • Provides phase contrast viewing.
  • Small.
  • Insertable.
  • Works with U-UCD8 universal condenser.
Specifications
Specifications
PlanN100XOPh, UPlanFLN60XOPh, UPlanFLN100XOPh
Small
Phase contrast ring