missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-4,5-dicarboxylic acid, 97%, Thermo Scientific™

Product Code. 15491078
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15491078

Brand: Thermo Scientific Alfa Aesar H54939.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5462-82-8
Molecular Formula C16H10O4
Molecular Weight (g/mol) 266.252
MDL Number MFCD11110581
InChI Key RAGXPRCIICFJIK-UHFFFAOYSA-N
Synonym 4,5-phenanthrenedicarboxylic acid, phenanthrene-4,5-dicarboxylate, 4,5-phenanthrenedicarboxylicacid, phenanthrene-4,5-dicarboxylic acid
PubChem CID 95075
ChEBI CHEBI:81603
IUPAC Name phenanthrene-4,5-dicarboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C3=C(C=CC=C3C(=O)O)C=C2

Specifications

Melting Point 244°C to 246°C
Quantity 250 mg
Formula Weight 266.25
Percent Purity 97%
Chemical Name or Material Phenanthrene-4,5-dicarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.