missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phenol Red, ACS

pH indicator

Brand:  Thermo Scientific™ 016294.22

Product Code. 15461077

  • 194.00€ / 100g
 Restricted Item
Estimated Shipment: 15-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

143-74-8
MFCD00003552
14,7247
Practically insoluble in chloroform,ether; Soluble in aqueous alkali hydroxides or carbonates with red color (removed by boiling with zinc dust); Solubility in water: approximately 1g/1300ml; Solubility in alcohol: approximately 1g/350mlSoluble in methanol.
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
ACS
100g
C19H14O5S
326470
Phenolsulfonphthalein; Phenylsulfonephthalein
BELBBZDIHDAJOR-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
4766
354.38
Odorless
Phenol Red
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.