Learn More
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
Product Code. 11883825
Change view
Click to view available options
Liner Material:
Rubber
Unit Size:
Pack of 1000
Product Code. Liner Material unitSize
11883825 Rubber Pack of 1000
11868532 Rubber Pack of 1000
2 options
Product Code. 11883825
Brand: DWK Life Sciences 7380015415
Only null left
Add to basket
Edge
Add to basket
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
  • Styrene Liner
  • Screw Cap
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 15-415
Liner Material Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Description Fixed Screw Cap
Material Phenolic
Color Black
For Use With (Application) For general laboratory use
Quantity 1000 Pack