Learn More
Phenolophtalein 20 mg/L in Propanol-2 Indicator Solution, Chem Lab
Brand: Chem Lab CL06.0907.9010
Additional Details : Weight : 7.80000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Specifications
-89°C | |
Polyethylene | |
Colorless | |
12 °C |
82°C | |
0.78 g/mol | |
10 L | |
Liquid |
Safety and Handling
- Phenolphtalein 20 mg/l in propanol-2 indicator solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol