missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphtalein Indicator in Ethanol, 2g/L, Chem-Lab
Click to view available options
Quantity:
1 L
Description
Phenolphthalein is usually dissolved in alcohol for use in experiments. It is a weak acid, which can lose H+ ions in solution. The phenolphthalein molecule is colorless, while the phenolphthalein ion is pink. In acidic and neutral solutions, the solution is colorless. When a base is added to the phenolphthalein, more ionization takes place, causing the solution to turn a red-violet color. This solution turns this pinkish color at pH 8.2–9.8.
Specifications
Melting Point | -114°C |
CAS | 77-09-8, 64-17-5 |
Concentration | 2g C20H14O4 per 1L C2H5OH |
Molecular Formula | C20H14O4 |
MDL Number | MFCD00005913 |
Solubility Information | Slightly soluble in water |
InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
IUPAC Name | 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
Molecular Weight (g/mol) | 318.33 |
Show More |
Safety and Handling
Product Identifier
- Phenolphtalein indicator in Ethanol
- Danger
- Flammable liquid Category 2
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- H225-Highly flammable liquid and vapour.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
- H361-Suspected of damaging fertility or the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'