Learn More
Macherey Nagel Phenolphthalein Test Paper, Refill
pH indicator paper for phenolphthalein, without color scale, refill.
Brand: Macherey Nagel 90713
This item is not returnable.
View return policy
Description
- Simplest acid / base determination
- Qualitative pH papers without color chart are simple, completely impregnated indicator papers.
- They indicate if the pH of a solution is above or below the transition point (color change) and are useful to distinguish between acids and bases.
Specifications
| White to Red/pH 8.3 to 10.0 | |
| 8.3 to 10 | |
| 5 m | |
| 7 mm |