Learn More
Macherey Nagel Phenolphthalein Test Paper, Refill
pH indicator paper for phenolphthalein, without color scale, refill.
Brand:  Macherey Nagel 90713
Product Code. 11316524
  • ONLINE EXCLUSIVE PRICE
    17.45€
    16.80€ / Pack of 2
    Save 0.65€
    3% Off
Only null left
In Stock
Add to basket
Description
Description
  • Simplest acid / base determination
    • Qualitative pH papers without color chart are simple, completely impregnated indicator papers.
    • They indicate if the pH of a solution is above or below the transition point (color change) and are useful to distinguish between acids and bases.
Specifications
Specifications
White to Red/pH 8.3 to 10.0
8.3 to 10
5 m
7 mm