Learn More
Phenoxybenzamine hydrochloride, Thermo Scientific™
30.10€ - 47.50€
Chemical Identifiers
| CAS | 63-92-3 |
|---|---|
| Molecular Formula | C18H23Cl2NO |
| Molecular Weight (g/mol) | 340.288 |
| MDL Number | MFCD00055152 |
| InChI Key | VBCPVIWPDJVHAN-UHFFFAOYSA-N |
| Synonym | phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine |
| PubChem CID | 5284441 |
| ChEBI | CHEBI:8078 |
| IUPAC Name | N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride |
| SMILES | CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15454389
|
Thermo Scientific Alfa Aesar
J61900.MD |
250mg |
30.10€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15444389
|
Thermo Scientific Alfa Aesar
J61900.03 |
47.50€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Selective alpha adrenergic blocking agent
- Irreversible calmodulin inhibitor
Chemical Identifiers
| 63-92-3 | |
| 340.288 | |
| VBCPVIWPDJVHAN-UHFFFAOYSA-N | |
| 5284441 | |
| N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride |
| C18H23Cl2NO | |
| MFCD00055152 | |
| phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine | |
| CHEBI:8078 | |
| CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl |
Specifications
| 63-92-3 | |
| MFCD00055152 | |
| phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine | |
| CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl | |
| 340.288 | |
| CHEBI:8078 | |
| Phenoxybenzamine hydrochloride |
| C18H23Cl2NO | |
| 14,7256 | |
| VBCPVIWPDJVHAN-UHFFFAOYSA-N | |
| N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride | |
| 5284441 | |
| 340.29 |
Safety and Handling
GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
missing translation for 'einecsNumber' : 200-569-7
missing translation for 'rtecsNumber' : DP3750000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.