missing translation for 'onlineSavingsMsg'
Learn More

Phenyl(4-piperidyl)methanone hydrochloride, 97%, Thermo Scientific™

Catalog Number p-4510486
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
25519-80-6
C12H16ClNO
225.716
NXYKIFZJQXOUJS-UHFFFAOYSA-N
4-benzoylpiperidine hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, 4-benzoylpiperidine hcl, phenyl 4-piperidyl methanone hydrochloride, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, 4-benzoyl piperidine hcl, pubchem9881
2724437
phenyl(piperidin-4-yl)methanone;hydrochloride
C1CNCCC1C(=O)C2=CC=CC=C2.Cl
This item is not returnable. View return policy
25519-80-6
C12H16ClNO
225.716
NXYKIFZJQXOUJS-UHFFFAOYSA-N
4-benzoylpiperidine hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, 4-benzoylpiperidine hcl, phenyl 4-piperidyl methanone hydrochloride, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, 4-benzoyl piperidine hcl, pubchem9881
2724437
phenyl(piperidin-4-yl)methanone;hydrochloride
C1CNCCC1C(=O)C2=CC=CC=C2.Cl
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 225.72
Percent Purity 97%
Chemical Name or Material Phenyl(4-piperidyl)methanone hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'