missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™

Catalog Number p-181945
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
101-48-4
166.22
MFCD00008487
WNJSKZBEWNVKGU-UHFFFAOYSA-N
2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
60995
2,2-dimethoxyethylbenzene
COC(CC1=CC=CC=C1)OC
This item is not returnable. View return policy
101-48-4
166.22
MFCD00008487
WNJSKZBEWNVKGU-UHFFFAOYSA-N
2,2-dimethoxyethyl benzene, phenylacetaldehyde dimethyl acetal, viridine, hyscylene p, 1,1-dimethoxy-2-phenylethane, benzene, 2,2-dimethoxyethyl, phenacetaldehyde dimethyl acetal, padma, 2,2-dimethoxy-1-phenylethane, phenylacetic aldehyde dimethyl acetal
60995
2,2-dimethoxyethylbenzene
COC(CC1=CC=CC=C1)OC
Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 1.0040g/mL
Boiling Point 219°C to 221°C (754.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O2
Linear Formula C6H5CH2CH(OCH3)2
Refractive Index 1.4930 to 1.495
Quantity 5 g
Beilstein 07, 293
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 2 parts of 70% alcohol
Formula Weight 166.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde dimethyl acetal, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'