Learn More
Knick Elektronische™ pH meter Portavo 904 digital
Includes datalogger and print functionalities. Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
Brand:  Knick Elektronische™ 904 pH
Additional Details : Weight : 0.56000kg
Product Code. 10089360
  • 1489.30€ / Each
Estimated Shipment: 11-04-2024
to see stock.
Récupel-bijdrage uitgesloten
Description
Description
  • pH Advanced pH Meter with datalogger and print functionalities includes remote-control capability.
  • Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
  • 15mm character height LC display with antireflection natural glass is scratchproof and resistant to chemicals.
  • Clear icons for operator guidance.
  • Integrated, replaceable quiver protects the sensor. Transparent. Easy to remove.
  • Blue, ergonomic enclosure is resistant to impact and chemicals. Water-tight, diecast chassis. (IP 66 protection).
  • Cover protects against dirt and damage.
  • Highly flexible lamellar joint can be folded back completely.
  • Fold-out for hanging or standing up keeps both hands free for handling the sensor.
  • Robust, gold-plated sockets are water-tight.
  • Adjustable carrying strap is fixed directly to the meter.
  • 1-, 2-, and 3-point calibration.
  • Calimatic™ automatically recognizes the right buffer fro 8 buffer sets.
  • Manual calibration possible with any two buffers.
  • Automatic recording of measured values over longer periods. Manual (with STO key), interval (e. g. every 30 min.) or eventcontrolled (based on a measured value difference – this function prevents unnecessary memory consumption).
  • Combined with a PC, the meter can continuously record any amount of data. Either for printer or PC simply by turning plug around. Saves paperwork, prevents manual transcription errors. Supplies records for QM documentation (ISO 9000 and GLP).
  • Paraly™ SW 109 transfer software transfers measured values to the PC.
Specifications
Specifications
With clock, measured value memory, data logger, interface, transfer software
904 digital
pH-Meter