Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), RNase free
Product Code. 15480267
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15480267
Brand: Thermo Scientific Chemicals J62851.AK
Request Bulk or Custom Format
Helps to maintain a constant pH
Phosphate-buffered saline (PBS, 10X), is an isotonic, non-toxic to most cells and is used in biological research. Phosphate-buffered saline with EDTA is used to disengage attached and clumped cells. It is also used in cell culturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
Physical Form Liquid
Quantity 250 mL