missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-S1P1 (Thr236) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540299
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Phospho-S1P1 (Thr236) Polyclonal antibody specifically detects Phospho-S1P1 (Thr236) in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Phospho-S1P1 (Thr236) | |
Polyclonal | |
Unconjugated | |
S1PR1 | |
AI849002; CD antigen CD363; CD363; CD363 antigen; CHEDG1; D1S3362; ECGF1; EDG1; EDG-1; EDG1 (Edg1); Edg1; rhodopsin family G protein-coupled receptor; endothelial differentiation G-protein coupled receptor 1; endothelial differentiation sphingolipid G-protein-coupled receptor 1; endothelial differentiation, sphingolipid G-protein-coupled receptor, 1; etID43307.22; Lpb1; Lysophospholipid receptor B1; S1p; S1P receptor 1; S1P receptor Edg-1; s1p1; S1PR1; sphingosine 1-phosphate receptor; sphingosine 1-phosphate receptor 1; sphingosine 1-phosphate receptor EDG1; Sphingosine 1-phosphate receptor Edg-1; sphingosine-1-phosphate receptor 1; wu:fb98f05 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13609, 1901, 29733 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08530, P21453, P48303 | |
S1PR1 | |
Synthesized peptide derived from human EDG-1 around the phosphorylation site of T236. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |