missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Leptin Receptor (Tyr1141) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540301
Additional Details : Weight : 0.00910kg
Description
This antibody is predicted to react with mouse based on sequence homology.
Phospho-Leptin Receptor (Tyr1141) Polyclonal antibody specifically detects Phospho-Leptin Receptor (Tyr1141) in Human, Mouse samples. It is validated for ELISA, Western Blot.Specifications
Phospho-Leptin Receptor (Tyr1141) | |
Polyclonal | |
Unconjugated | |
Lepr | |
B219; CD antigen CD295; CD295; CD295 antigen; db; diabetes; Fa; HuB219; HuB219.2; LEPR; LEP-R; Leprb; LEPRD; LEPROT; leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; Modb1; OB receptor; obese-like; obl; Obr; OB-R; OB-RGRP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16847, 3953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48356, P48357 | |
Lepr | |
Synthesized peptide derived from human Ob-R around the phosphorylation site of Y1141. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |