missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MYL9 (Thr18, Ser19) Recombinant Rabbit Monoclonal Antibody (18H3L4), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen 702368

Additional Details : Weight : 0.01000kg

Product Code. 15607748

  • 457.00€ / 100µg
Estimated Shipment: 15-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is predicted to react with Monkey, Cat and Pig Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Phospho-MYL9 (Thr18, Ser19) Recombinant Monoclonal antibody specifically detects Phospho-MYL9 (Thr18, Ser19) in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

Phospho-MYL9 (Thr18, Ser19)
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P24844, Q9CQ19
MYL9
Peptide corresponding to Human MYL9 (aa 14-23)
100 μg
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Western Blot
18H3L4
Unconjugated
MYL9
20 kDa myosin light chain; AI327049; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; myl9; myl9.L; myl9b; myl9l; Mylc2c; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light polypeptide 9; myosin regulatory light polypeptide 9b; myosin RLC; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; RLC-C; smooth muscle major isoform; unnamed protein product; XELAEV_18003093mg; zgc:77916
Rabbit
Protein A
RUO
10398, 98932
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions