missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus Pentachloride, Extra Pure, SLR, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Packaging:
Plastic Coated Glass bottle
Unit Size:
250g
Specifications
Specifications
| Boiling Point | 160°C |
| Melting Point | 167°C |
| CAS | 10026-13-8 |
| Color | White |
| pH | 1 |
| Physical Form | Powder/Solid |
| Quantity | 250 g |
| Molecular Formula | Cl5P |
| MDL Number | 11439 |
| Synonym | phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu |
| Show More |
Safety and Handling
Product Identifier
- Phosphorus pentachloride
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction