missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Standard: P @ 5000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17942146
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942146 50 g 50g
17971186 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942146 Supplier VHG Labs Supplier No. VHGOP500050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Phosphorus Standard: P @ 5000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus
CAS 7723-14-0
Methods ICP/XRF/RDE
Analyte or Component Names Phosphorus
Shipping Condition Room Temperature
Molecular Formula P
Certifications/Compliance ISO 17034
Synonym P
InChI Formula InChI=1S/P
SMILES [P]
IUPAC Name phosphorus
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 30.97
Quantity 50 g
Formula Weight 30.9738 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.