missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus trichloride, 99%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 250 g |
| Assay Percent Range | 99% |
| Molecular Formula | Cl3P |
| Linear Formula | PCl3 |
| MDL Number | MFCD00011438 |
| Fieser | 01,875; 08,400; 10,318 |
| InChI Key | FAIAAWCVCHQXDN-UHFFFAOYSA-N |
| SMILES | P(Cl)(Cl)Cl |
| IUPAC Name | trichlorophosphane |
| Show More |
Safety and Handling
Product Identifier
- Phosphorus trichloride
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction