missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus trichloride, 99%, Thermo Scientific Chemicals

Product Code. 10542191 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10542191

Brand: Thermo Scientific Acros 169480010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7719-12-2
Synonym phosphorus chloride pcl3, phosphorous trichloride, phosphorus trichloride, phosphorus iii chloride, fosfortrichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, pcl3, phosphine, trichloro
PubChem CID 24387
ChEBI CHEBI:30334

Specifications

Physical Form Liquid
Quantity 1 kg
Assay Percent Range 99%
Molecular Formula Cl3P
Linear Formula PCl3
MDL Number MFCD00011438
Fieser 01,875; 08,400; 10,318
InChI Key FAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILES ClP(Cl)Cl
IUPAC Name trichlorophosphane
Molecular Weight (g/mol) 137.32
Formula Weight 137.33
Percent Purity ≥98.5%
Packaging Glass bottle
Density 1.5700 g/mL
Chemical Name or Material Phosphorus trichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus trichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.