Learn More
Thermo Scientific™ Phusion Hot Start II High-Fidelity PCR Master Mix
Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR.
221.00€ - 872.00€
Specifications
Content And Storage | • 2X Phusion Hot Start II High-Fidelity PCR Master Mix • Nuclease-free water • DMSO 2X Phusion Hot Start II High-Fidelity PCR Master Mix provides 1.5 mM MgCl2 in the final 1X concentration. Store at –20°C. |
---|---|
GC-Rich PCR Performance | Medium |
Polymerase | Phusion Hot Start II High-Fidelity DNA Polymerase |
Reaction Speed | Fast |
Product Type | Hot Start High-Fidelity PCR Master Mix |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15301732
|
Thermo Scientific™
F565S |
100 Reactions |
221.00€
100 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15311732
|
Thermo Scientific™
F565L |
500 Reactions |
872.00€
500 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.
Highlights
• Reaction set up at room temperature
• No non-specific amplification and primer degradation during reaction set up
• Zero-time reactivation due to unique hot start technology
• High fidelity (52X Taq)
• Fast PCR due to short extension times (15-30 s/kb)
• Robust reactions, minimal optimization needed
• Increased product yields with minimal enzyme amounts
• Available as a Green buffer format permitting direct loading on gels (F-566S or F-566L)
Using Phusion Hot Start II High-Fidelity DNA Polymerase, amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3´→5´ exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols. Phusion Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. Only template and primers need to be added to the PCR reaction.
Applications
• High-fidelity PCR
• High throughput
• Amplification of difficult (GC-rich) templates
• Template generation for sequencing
• Multiplex PCR
• Long-range PCR
• Cloning
• Mutagenesis
• Microarray
Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our Tm calculator at www.thermofisher.com/tmcalculator.
Specifications
• 2X Phusion Hot Start II High-Fidelity PCR Master Mix • Nuclease-free water • DMSO 2X Phusion Hot Start II High-Fidelity PCR Master Mix provides 1.5 mM MgCl2 in the final 1X concentration. Store at –20°C. |
|
Phusion Hot Start II High-Fidelity DNA Polymerase | |
Hot Start High-Fidelity PCR Master Mix | |
Hot-start PCR, High-fidelity PCR | |
52X | |
Blunt | |
20 kb or less |
Medium | |
Fast | |
Dry Ice | |
2X | |
Built-In Hot Start | |
SuperMix or Master Mix |
For Research Use Only. Not for use in diagnostic procedures.