missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic X-Foam HV Safety Gloves

Product Code. 17250371 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17250371

Brand: uvex 6005411

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A seamless break section integrated into the fingers significantly reduces the risk of serious hand injuries when using manually operated screw drivers. Aqua-polymer foam coating on the palm and fingertips enhances grip capabilities.

  • Lightweight construction acts as a second-skin to maintain a high level of feeling
  • Aqua-polymer foam coating is moisture repellent and flexible
  • Breathable materials help prevent excessive perspiration
  • Seamless break section integrated into the fingers helps reduce serious hand injuries
  • Should the glove/finger become trapped in or near a piece of machinery, simply detach and tear along the perforated seam
  • Enhanced grip in dry and slightly damp applications
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Orange, Gray
Hand Ambidextrous
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 11
EN388 Standard 31X1X
Gender Unisex
Glove Cuff Style Knit
Series Phynomic X-Foam HV
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.