Learn More
Eurofins DiscoverX PI3 Kinase (p110β/p85β) Mouse Protein
Description
Phosphoinositide 3-kinases (PI3Ks) phosphorylate the 3-prime OH position of the inositol ring of inositol lipids. They have been implicated as participants in signaling pathways regulating cell growth by virtue of their activation in response to various mitogenic stimuli.
GenBank NM_029094. The recombinant protein contains the amino acid substitution R123L (native protein coordinates) with respect toGenBank NM_029094, this is reported in the ESTs CV558291, CO431848, CN701015, CN681036, CN536135 and CN534564 / GenBank BC006796
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 125.5 kDa (p110β) and 81.4 kDa (p85β) |
| Quantity | 10 μg |
| Species | Murine |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / Untagged |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥72% |
| Protein | PI 3-Kinase (p110β/p85β), murine |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.