Learn More
Pierce™ 20X Tris-HEPES-SDS Buffer, Thermo Scientific™
Product Code. 10230064 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 10230064
Brand: Thermo Scientific 28368
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Prepare Tris-HEPES-SDS running buffer required for gel electrophoresis (SDS-PAGE) with Precise Precast Gels with this space-saving stock solution.
Thermo Scientific™ Pierce 20X Tris-HEPES-SDS Buffer is a space-saving stock solution used for preparing the Tris-HEPES-SDS running buffer required for gel electrophoresis (SDS-PAGE) with Thermo Scientific™ Precise and Pierce Precast Gels.
Thermo Scientific Pierce 10X and 20X Concentrated Buffers are ready to use without having to reconstitute with ultrapure water.
Highlights:
  • Easy to use – no packets to open and no powder to dissolve
  • Increased accuracy – eliminates the possibility of powder remaining in a packet
  • Saves time – 10X or 20X concentration eliminates time spent waiting for powder to dissolve
  • Saves space – storage as concentrated stock minimizes bench space needed for large volume solutions
Ordering Alerts:
When diluted 20-fold in water, makes 0.1M Tris, 0.1M HEPES, 3mM SDS, pH 8
Recommended for:
Running buffer for SDS-PAGE with Thermo Scientific Precise™ and Pierce™ Precast Gels
TRUSTED_SUSTAINABILITY
Specifications
Description Pierce 20X Tris-HEPES-SDS Buffer
For Use With (Application) Electrophoresis
Quantity 500 mL
Formulation 1X = 0.1M Tris, 0.1M HEPES, 3mM SDS, pH 8