Learn More
Thermo Scientific™ Pierce™ DTT (Dithiothreitol), No-Weigh™ Format
Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.
POA
Description
Description
Thermo Scientific™ Pierce DTT (Dithiothreitol) is offered as 7.7mg × 48-tube microplates for convenient preparation of 100μL of 0.5M stocks for addition to protein samples for effective reduction of disulfide bonds.
Highlights:
  • A water-soluble reagent that reduces disulfide bonds
  • Maintains mono-thiols completely in the reduced state and reduces disulfide bonds quantitatively
  • Specific and sensitive assay for disulfides using DTT with Ellman's Reagent