Learn More
Invitrogen™ PIP4K2A Recombinant Human Protein
This PIP4K2A Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A33519
548.06 EUR valid until 2023-03-27
Use promo code "20585" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- This PIP4K2A recombinant human protein (full length) was expressed in insect cells
- PIP4K2A plays as essential role in the phosphoinositide signal transduction cascades as the precursor to second messengers
- It is involved in the regulation of secretion, cell proliferation, differentiation, and motility
- PIP4K2A may be one of the factors related to the regulation of the beta-globin gene expression and the different levels of Hb H in alpha-thalassemic patients
- PIP4K2A recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
- Also available in our SelectScreen Kinase Profiling Service
Specifications
NP_005019.2 | |
20 µg | |
Full Length | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Human |
72.5 kDa | |
PI3 Kinase | |
Yes | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Baculovirus | |
Full Length, Recombinant, Active | |
Kinase Assay |