missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIP5K1B, GST Tag Recombinant Protein

Product Code. 17128855
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
Product Code. Quantity unitSize
17128855 1 mg 1mg
15940423 20 μg 20µg
17178825 100 μg 100µg
3 options
This item is not returnable. View return policy

Product Code. 17128855

Brand: Invitrogen™ A33741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIP5K1B recombinant human protein (full length) was expressed in insect cells. PIP5K1B recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIP5K1B is NP_003549.1.

PIP5K1B lipid kinase or phosphatidylinositol-4-phosphate 5-kinase, type I, beta is a member of the phosphatidylinositol-4-phosphate 5-kinase family. PIP5K1B gene contains 17 exons and spans more than 300 kb. The seventeenth exon was found by RT-PCR, and is derived from the 3-prime untranslated region of the PRKACG gene which is located on 9q13 approximately 3 kb downstream of the STM7.I 3-prime untranslated region. The overexpression of PIP5K1B in COS-7 cells induces an increase in short actin fibers and a decrease in actin stress fibers.
  • Validated for use with the Adapta™ assay
  • Available in multiple pack sizes for convenient use
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_003549.1
For Use With (Application) Kinase Assay
Molecular Weight (g/mol) 87.3 kDa
Quantity 1 mg
KinaseFamily PI3 Kinase
Protein Length Full Length
Species Human
Recombinant Recombinant
Protein Tag GST Tag
Content And Storage Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol
Shipping Condition Dry Ice
Expression System Insect
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.