missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIP5K1C, GST Tag Recombinant Protein

Product Code. 17138855
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138855 1 mg 1mg
15950423 20 μg 20µg
17188825 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17138855 Supplier Invitrogen™ Supplier No. A33742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIP5K1C recombinant human protein (full length) was expressed in insect cells. PIP5K1C recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIP5K1C is NP_036530.1.

PIP5K1C lipid kinase or phosphatidylinositol-4-phosphate 5-kinase 1 gamma phosphorylates phosphatidylinositol-4-phosphate to form phosphatidylinositol 4,5-bisphosphate. PIP5K1C is found at synapses and has been found to play roles in endocytosis and cell migration. PIP5K1C directly associates with β-catenin and increases β-catenin activity downstream of growth factor stimulation. PIP5K1C expression and kinase activity enhance β-catenin phosphorylation on residues that promote nuclear importation and transcriptional activity. PIP5K1C deficiency impairs neutrophil recruitment as a result of defect in adhesion. PIP5K1C regulated the adhesion through facilitation of RhoA GTPase and integrin activation by chemoattractants.
  • Validated for use with the Adapta™ assay
  • Available in multiple pack sizes for convenient use
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_036530.1
For Use With (Application) Kinase Assay
Molecular Weight (g/mol) 99.6 kDa
Quantity 1 mg
KinaseFamily PI3 Kinase
Protein Length Full Length
Species Human
Recombinant Recombinant
Protein Tag GST Tag
Content And Storage Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol
Shipping Condition Dry Ice
Expression System Insect
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.