missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pipemarker App (CLP-Rollform) for Brady Workstation

Product Code. 30281108
Click to view available options
:
Pipemarker App
This item is not returnable. View return policy

Product Code. 30281108

Brand: Brady™ 149205

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

With the Pipemarker (CLP-Rollform) App for Brady Workstation, with it's predefined templates and rollform layout, you can create customized, CLP-compliant pipe markers to wrap around your pipe.

  • For Use With (Equipment): BBP30, BBP31, BBP33, BBP35, BBP37, BBP85, BMP71, IP Series, J2000, J4000, J5000, i3300, S3000, GlobalMark 2 Multicolor

Specifications

Product Type Software
For Use With (Equipment) Brady Label Printers
Format Email with Activation Code
Languages Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish (Mexico), Spanish, Swedish, Turkish, Vietnamese
Software Type Sign and Label Design Software
Alternate Part Number BWS-RLF-EM
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.