Learn More
PIPES lysis buffer with Triton™ X-100, Thermo Scientific™
Product Code. 15404759
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15404759
Brand: Thermo Scientific Alfa Aesar J62360.AK
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material PIPES lysis buffer with Triton™ X-100
Physical Form Liquid
Quantity 250 mL