Learn More
ratiolab™ Pipet Tips
Click to view available options
Color:
Blue
Clear
Yellow
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
1 to 250 μL
100 to 1000 μL
100 to 1200 μL
500 to 2500 μL
Packaging:
Bag
Multirack
Unit Size:
Pack of 1000
Pack of 250
Pack of 600
Pack of 960
Description
- Guaranteed dust-free due to special ionization treatment
- High transparency due to high quality surfaces
- Minimal residual wetting due to the controlled quality of raw materials
- High volume accuracy due to precise tip orifice
- Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Specifications
Specifications
| Volume (Metric) | 1 to 250 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Packaging | Bag |
| Quantity | 1000 Tips |
| Material | Polypropylene |