Learn More
ratiolab™ Pipet Tips
Product Code. 11303310
Click to view available options
Color:
Blue
Clear
Yellow
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
1 to 250 μL
100 to 1000 μL
100 to 1200 μL
500 to 2500 μL
Packaging:
Bag
Multirack
Unit Size:
Pack of 1000
Pack of 250
Pack of 600
Pack of 960
Product Code. 11303310
Brand: ratiolab™ 2500163
Only null left
Add to basket
Add to basket
Pipet tips for all commercially available manual or electronic, single or multichannel pipets.
  • Guaranteed dust-free due to special ionization treatment
  • High transparency due to high quality surfaces
  • Minimal residual wetting due to the controlled quality of raw materials
  • High volume accuracy due to precise tip orifice
  • Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Specifications
Volume (Metric) 1 to 250 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Packaging Bag
Quantity 1000 Tips
Material Polypropylene